siRNA Details
| Gene Symbol: Snx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080557.2 | NP_542124.1 | 6, 7 | 669 | ||
| GenBank mRNA | AK007706.1 | - | 6, 7 | 711 | ||
| AK030657.1 | - | 6, 7 | 682 | |||
| AK031368.1 | - | 681 | ||||
| AK135356.1 | - | 6, 7 | 676 | |||
| BC006043.1 | - | 6, 7 | 659 | |||
| BC016599.1 | - | 6, 7 | 669 | 2644991 | ||

Snx4
Genomic Map