siRNA Details
| Gene Symbol: Krtap6-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130856.2 | NP_570926.1 | 1 | 446 | ||
| GenBank mRNA | AF345298.1 | - | 1 | 428 | ||
| AK003281.1 | - | 1 | 447 | |||
| BC059086.1 | - | 1 | 430 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap6-5| Gene Symbol: Krtap6-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130856.2 | NP_570926.1 | 1 | 446 | ||
| GenBank mRNA | AF345298.1 | - | 1 | 428 | ||
| AK003281.1 | - | 1 | 447 | |||
| BC059086.1 | - | 1 | 430 | |||