siRNA Details
| Gene Symbol: Dazap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122604.1 | NP_001116076.1 | 3, 4 | 388 | ||
| NM_001122605.1 | NP_001116077.1 | 3, 4 | 388 | |||
| NM_133188.2 | NP_573451.2 | 3, 4 | 388 | |||
| GenBank mRNA | AF225910.1 | - | 3, 4 | 282 | ||
| AK148023.1 | - | 3, 4 | 266 | |||
| AK152314.1 | - | 3, 4 | 358 | |||
| AK152345.1 | - | 3, 4 | 358 | |||
| BC049355.1 | - | 3, 4 | 305 | 6337388 | ||
| BC058775.1 | - | 3, 4 | 292 | |||
| CJ171666.1 | - | 3, 4 | 393 | |||

Dazap1
Genomic Map