siRNA Details
| Gene Symbol: Lyrm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163628.1 | NP_001157100.1 | 3 | 814 | ||
| NM_133688.3 | NP_598449.1 | 3 | 577 | |||
| GenBank mRNA | AF412298.1 | - | 3 | 1231 | ||
| AF412299.1 | - | 4 | 812 | |||
| AF412300.1 | - | 3 | 578 | |||
| AK015530.1 | - | 3 | 550 | |||
| AY753325.1 | - | 3 | 569 | |||
| BC021522.1 | - | 3 | 525 | 5352723 | ||

Lyrm5
Genomic Map