siRNA Details
| Gene Symbol: Rbm42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133693.2 | NP_598454.2 | 9 | 1332 | ||
| GenBank mRNA | AK013929.1 | - | 9 | 1273 | ||
| AK151000.1 | - | 9 | 1244 | |||
| AK189364.1 | - | 1 | 33 | |||
| AK215240.1 | - | 1 | 33 | |||
| BC009148.1 | - | 9 | 1269 | 3600406 | ||
| BC011286.1 | - | 9 | 1251 | 3485410 | ||
| BC027372.1 | - | 9 | 1256 | 4952304 | ||
| BC039917.1 | - | 9 | 1267 | |||
| BC057928.1 | - | 9 | 1258 | 5289543 | ||
| BY551742.1 | - | 2 | 132 | |||

Rbm42
Genomic Map