siRNA Details
| Gene Symbol: Btbd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133700.2 | NP_598461.1 | 5, 6 | 659 | ||
| GenBank mRNA | BC050781.1 | - | 5, 6 | 659 | 6591397 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd10| Gene Symbol: Btbd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133700.2 | NP_598461.1 | 5, 6 | 659 | ||
| GenBank mRNA | BC050781.1 | - | 5, 6 | 659 | 6591397 | |