siRNA Details
| Gene Symbol: Tmem97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133706.2 | NP_598467.1 | 3 | 416 | ||
| GenBank mRNA | AK007497.1 | - | 2 | 145 | ||
| AK155107.1 | - | 3 | 428 | |||
| AK171954.1 | - | 3 | 395 | |||
| BC018156.1 | - | 3 | 416 | 4192256 | ||
| BC093511.1 | - | 3 | 393 | 5124076 | ||

Tmem97
Genomic Map