siRNA Details
| Gene Symbol: Ctdspl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133710.3 | NP_598471.3 | 2 | 421 | ||
| GenBank mRNA | AA239542.1 | - | 2 | 112 | ||
| AJ344340.1 | - | 2 | 503 | |||
| AK013115.1 | - | 2 | 418 | |||
| BC094289.1 | - | 2 | 106 | 4986338 | ||

Ctdspl
Genomic Map