siRNA Details
| Gene Symbol: Spata4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133711.3 | NP_598472.2 | 5, 6 | 815 | ||
| GenBank mRNA | AF395083.2 | - | 5, 6 | 815 | ||
| AF530515.1 | - | 5, 6 | 815 | |||
| AK005612.1 | - | 5, 6 | 815 | |||
| BC048650.1 | - | 5, 6 | 815 | 6772684 | ||

Spata4
Genomic Map