siRNA Details
| Gene Symbol: Asnsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133728.3 | NP_598489.2 | 4 | 1171 | ||
| GenBank mRNA | AK008875.1 | - | 4 | 1087 | ||
| AK031295.1 | - | 4 | 1129 | |||
| AK034746.1 | - | 1090 | ||||
| AK049194.1 | - | 4 | 1118 | |||
| AK050045.1 | - | 4 | 1110 | |||
| AK076793.1 | - | 4 | 1073 | |||
| AK155030.1 | - | 4 | 1110 | |||
| AK168103.1 | - | 4 | 1111 | |||
| AK170510.1 | - | 4 | 1087 | |||
| BC013617.1 | - | 4 | 991 | 3979452 | ||

Asnsd1
Genomic Map