siRNA Details
| Gene Symbol: Stk26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133729.1 | NP_598490.1 | 11 | 1335 | ||
| GenBank mRNA | AK011445.1 | - | 11 | 1719 | ||
| AK029278.1 | - | 9 | 1940 | |||
| AK034609.1 | - | 11 | 1374 | |||
| BC005708.1 | - | 11 | 1335 | 2598933 | ||
| BC132438.1 | - | 10 | 1138 | 40130715 | ||
| BC138193.1 | - | 10 | 1138 | 8861212 | ||
| BC145189.1 | - | 10 | 1155 | |||

Stk26
Genomic Map