siRNA Details
| Gene Symbol: Hemk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088071.1 | - | 2 | 830 | ||
| BC011431.1 | - | 2 | 470 | 3981356 | ||
| BC024428.1 | - | 2 | 330 | 4971671 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hemk1| Gene Symbol: Hemk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088071.1 | - | 2 | 830 | ||
| BC011431.1 | - | 2 | 470 | 3981356 | ||
| BC024428.1 | - | 2 | 330 | 4971671 | ||