siRNA Details
| Gene Symbol: Reep6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204931.1 | NP_001191860.1 | 3 | 399 | ||
| NM_139292.2 | NP_647453.1 | 3 | 399 | |||
| GenBank mRNA | AB039933.1 | - | 3 | 374 | ||
| AK002562.1 | - | 3 | 397 | |||
| AY562234.1 | - | 3 | 313 | |||
| BC029741.1 | - | 3 | 386 | 4195889 | ||

Reep6
Genomic Map