siRNA Details
| Gene Symbol: Zfp639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161818.1 | NP_001155290.1 | 6 | 515 | ||
| NM_144519.4 | NP_653102.1 | 6 | 616 | |||
| GenBank mRNA | AK049934.1 | - | 6 | 495 | ||
| AK076145.1 | - | 6 | 617 | |||
| AK076837.1 | - | 6 | 492 | |||
| AK078000.1 | - | 6 | 516 | |||
| AY736186.1 | - | 3 | 201 | |||
| BC003941.1 | - | 4 | 265 | 3487151 | ||
| DQ323614.1 | - | 2 | 55 | |||

Zfp639
Genomic Map