siRNA Details
| Gene Symbol: Zfp639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161818.1 | NP_001155290.1 | 7 | 836 | ||
| NM_144519.4 | NP_653102.1 | 7 | 937 | |||
| GenBank mRNA | AK049934.1 | - | 7 | 816 | ||
| AK076145.1 | - | 7 | 938 | |||
| AK076837.1 | - | 7 | 813 | |||
| AK078000.1 | - | 7 | 837 | |||
| AK201027.1 | - | 1 | 112 | |||
| AK219010.1 | - | 1 | 33 | |||
| AY736186.1 | - | 4 | 522 | |||
| BC003941.1 | - | 5 | 586 | 3487151 | ||
| BC042660.1 | - | 1 | 178 | |||
| DQ323614.1 | - | 3 | 376 | |||

Zfp639
Genomic Map