siRNA Details
| Gene Symbol: Tbc1d10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144522.5 | NP_653105.3 | 2 | 1033 | ||
| GenBank mRNA | AF285112.1 | - | 2 | 391 | ||
| AK047320.1 | - | 2 | 1034 | |||
| AK047327.1 | - | 2 | 1034 | |||
| BC072576.1 | - | 2 | 576 | 6401332 | ||

Tbc1d10b
Genomic Map