siRNA Details
| Gene Symbol: Ncapd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146171.1 | NP_666283.1 | 2, 3 | 236 | ||
| GenBank mRNA | AK129068.1 | - | 2, 3 | 201 | ||
| AK156143.1 | - | 2, 3 | 266 | |||
| BC025460.1 | - | 2, 3 | 256 | |||
| BC026982.1 | - | 2, 3 | 236 | 5364933 | ||
| BC029133.1 | - | 2, 3 | 193 | 4951596 | ||

Ncapd2
Genomic Map