siRNA Details
| Gene Symbol: Zfp740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289690.1 | NP_001276619.1 | 6 | 2658 | ||
| NM_001289691.1 | NP_001276620.1 | 8 | 2857 | |||
| NM_001289694.1 | NP_001276623.1 | 7 | 2566 | |||
| NM_001289695.1 | NP_001276624.1 | 7 | 2532 | |||
| GenBank mRNA | AK144707.1 | - | 8 | 2855 | ||
| AK170207.1 | - | 7 | 2553 | |||
| BC030410.1 | - | 3 | 2363 | 5367159 | ||
| BC066011.1 | - | 7 | 2426 | 6418561 | ||

Zfp740
Genomic Map