siRNA Details
| Gene Symbol: Rnf113a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153503.1 | NP_705723.1 | 1 | 1059 | ||
| GenBank mRNA | AK013178.1 | - | 1 | 1120 | ||
| AK146283.1 | - | 1 | 1158 | |||
| BC024906.1 | - | 1 | 1059 | 3495576 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf113a1| Gene Symbol: Rnf113a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153503.1 | NP_705723.1 | 1 | 1059 | ||
| GenBank mRNA | AK013178.1 | - | 1 | 1120 | ||
| AK146283.1 | - | 1 | 1158 | |||
| BC024906.1 | - | 1 | 1059 | 3495576 | ||