siRNA Details
| Gene Symbol: Nrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153529.2 | NP_705757.1 | 2 | 366 | ||
| GenBank mRNA | AK003046.1 | - | 2 | 193 | ||
| AK161859.1 | - | 2 | 334 | |||
| BC035531.1 | - | 2 | 354 | 5367281 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053346, U88958, BC087582.

Nrn1
Genomic Map