siRNA Details
| Gene Symbol: Tra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB052758.1 | - | 1 | 361 | ||
| AK005187.1 | - | 1 | 1072 | |||
| AK076230.1 | - | 1 | 1058 | |||
| AK076606.1 | - | 1 | 1040 | |||
| AK148552.1 | - | 2 | 823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tra2a| Gene Symbol: Tra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB052758.1 | - | 1 | 361 | ||
| AK005187.1 | - | 1 | 1072 | |||
| AK076230.1 | - | 1 | 1058 | |||
| AK076606.1 | - | 1 | 1040 | |||
| AK148552.1 | - | 2 | 823 | |||