siRNA Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175121.3 | NP_780330.2 | 16 | 2532 | ||
| GenBank mRNA | AK033812.1 | - | 4 | 1073 | ||
| AK129342.1 | - | 16 | 2492 | |||
| AK164739.1 | - | 1 | 525 | |||
| AK169378.1 | - | 6 | 1290 | |||
| BC037675.1 | - | 13 | 1939 | |||
| BC041108.1 | - | 16 | 2402 | 4038546 | ||
| BC048178.1 | - | 16 | 2532 | 6825801 | ||
| BC049271.1 | - | 9 | 1549 | |||
| BC057454.1 | - | 3 | 913 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_181090, XM_006242283, XM_006242282, AF273024, AF249673.

Slc38a2
Genomic Map