siRNA Details
| Gene Symbol: Dnaic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175138.4 | NP_780347.2 | 7 | 775 | ||
| GenBank mRNA | AK030213.1 | - | 7 | 709 | ||
| AK047808.1 | - | 7 | 776 | |||
| BC138785.1 | - | 7 | 720 | 8861807 | ||
| BC138787.1 | - | 7 | 720 | 8861809 | ||

Dnaic1
Genomic Map