siRNA Details
| Gene Symbol: Chst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175140.4 | NP_780349.3 | 4 | 1647 | ||
| GenBank mRNA | AB106878.1 | - | 3 | 1245 | ||
| AF308145.1 | - | 4 | 1465 | |||
| AK005217.1 | - | 4 | 1381 | |||
| AK038952.1 | - | 3 | 1620 | |||
| AK051290.1 | - | 4 | 1229 | |||
| BC051124.1 | - | 4 | 1612 | 6528978 | ||

Chst8
Genomic Map