siRNA Details
| Gene Symbol: Tsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164578.1 | NP_001158050.1 | 5 | 814 | ||
| NM_175146.4 | NP_780355.2 | 5 | 805 | |||
| GenBank mRNA | AK044504.1 | - | 5 | 781 | ||
| BC025008.1 | - | 5 | 768 | |||
| BC037444.1 | - | 4 | 669 | |||
| BC049668.1 | - | 5 | 787 | 6535058 | ||

Tsr2
Genomic Map