siRNA Details
| Gene Symbol: 2300002M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175148.3 | NP_780357.1 | 2 | 1226 | ||
| GenBank mRNA | AK009013.1 | - | 2 | 1178 | ||
| AK037046.1 | - | 2 | 1229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2300002M23Rik| Gene Symbol: 2300002M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175148.3 | NP_780357.1 | 2 | 1226 | ||
| GenBank mRNA | AK009013.1 | - | 2 | 1178 | ||
| AK037046.1 | - | 2 | 1229 | |||