siRNA Details
| Gene Symbol: 2310022B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175149.4 | NP_780358.3 | 4 | 2398 | ||
| GenBank mRNA | AK084773.1 | - | 4 | 2219 | ||
| AK154813.1 | - | 4 | 2224 | |||
| BC058626.1 | - | 4 | 2398 | 6401577 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310022B05Rik| Gene Symbol: 2310022B05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175149.4 | NP_780358.3 | 4 | 2398 | ||
| GenBank mRNA | AK084773.1 | - | 4 | 2219 | ||
| AK154813.1 | - | 4 | 2224 | |||
| BC058626.1 | - | 4 | 2398 | 6401577 | ||