siRNA Details
| Gene Symbol: Thap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175152.4 | NP_780361.2 | 3, 4 | 404 | ||
| GenBank mRNA | AK008225.1 | - | 3, 4 | 376 | ||
| AK038184.1 | - | 3, 4 | 378 | |||
| AK050740.1 | - | 3, 4 | 418 | |||
| BC094395.1 | - | 3, 4 | 385 | 5700290 | ||
| BC115485.1 | - | 3, 4 | 402 | |||
| BC115486.1 | - | 3, 4 | 402 | |||

Thap3
Genomic Map