siRNA Details
| Gene Symbol: Vwa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077631.2 | NP_001071099.1 | 12, 13 | 1784 | ||
| NM_175153.4 | NP_780362.2 | 12, 13 | 2005 | |||
| GenBank mRNA | AK032095.1 | - | 12, 13 | 2000 | ||
| AK035654.1 | - | 12, 13 | 1712 | |||
| AK168222.1 | - | 11, 12 | 1687 | |||
| AK169772.1 | - | 12, 13 | 1744 | |||
| BC024913.1 | - | 11, 12 | 1689 | 3602181 | ||

Vwa9
Genomic Map