siRNA Details
| Gene Symbol: Vwa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077631.2 | NP_001071099.1 | 13 | 1792 | ||
| NM_175153.4 | NP_780362.2 | 13 | 2013 | |||
| GenBank mRNA | AK032095.1 | - | 13 | 2008 | ||
| AK035654.1 | - | 13 | 1720 | |||
| AK168222.1 | - | 12 | 1695 | |||
| AK169772.1 | - | 13 | 1752 | |||
| BC024913.1 | - | 12 | 1697 | 3602181 | ||

Vwa9
Genomic Map