siRNA Details
| Gene Symbol: Usp46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177561.3 | NP_808229.1 | 4 | 572 | ||
| GenBank mRNA | AK029604.1 | - | 3 | 424 | ||
| BC030888.1 | - | 2 | 339 | |||
| BC039916.1 | - | 4 | 574 | 4950034 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp46| Gene Symbol: Usp46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177561.3 | NP_808229.1 | 4 | 572 | ||
| GenBank mRNA | AK029604.1 | - | 3 | 424 | ||
| BC030888.1 | - | 2 | 339 | |||
| BC039916.1 | - | 4 | 574 | 4950034 | ||