siRNA Details
| Gene Symbol: Mterf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178051.3 | NP_835152.1 | 2 | 365 | ||
| GenBank mRNA | AK007901.1 | - | 2 | 352 | ||
| AK033043.1 | - | 2 | 376 | |||
| AK077142.1 | - | 2 | 365 | |||
| AK079098.1 | - | 2 | 357 | |||
| BC055877.1 | - | 1 | 309 | 4236729 | ||
| BC064709.1 | - | 2 | 358 | 30314353 | ||
| BY261356.1 | - | 2 | 371 | |||

Mterf4
Genomic Map