siRNA Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178065.4 | NP_835166.3 | 7 | 1433 | ||
| GenBank mRNA | AB093230.1 | - | 6 | 1052 | ||
| AK077015.1 | - | 6 | 965 | |||
| AK133332.1 | - | 6 | 968 | |||
| AK147491.1 | - | 7 | 1434 | |||
| AK147878.1 | - | 7 | 1397 | |||
| AK154529.1 | - | 7 | 1414 | |||
| BC049900.1 | - | 1 | 123 | |||
| BC057351.1 | - | 7 | 1371 | |||
| BC060658.1 | - | 6 | 1002 | 30355406 | ||
| BC079590.1 | - | 7 | 1391 | |||

Arel1
Genomic Map