siRNA Details
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011899.1 | - | 2, 3 | 220 | ||
AK082940.1 | - | 2, 3 | 240 | |||
AK167383.1 | - | 2, 3 | 228 | |||
AK167848.1 | - | 2, 3 | 224 | |||
AY082332.1 | - | 2, 3 | 194 | |||
BC050206.1 | - | 2, 3 | 202 | 30009463 |

More Information
Other Species Targeted

Panther Classification:
