siRNA Details
| Gene Symbol: Samm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031792.1 | - | 3296 | |||
| AK041783.1 | - | 2175 | ||||
| AK086633.1 | - | 2177 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samm50| Gene Symbol: Samm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031792.1 | - | 3296 | |||
| AK041783.1 | - | 2175 | ||||
| AK086633.1 | - | 2177 | ||||