siRNA Details
| Gene Symbol: RGS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006480.4 | NP_006471.2 | 12, 13 | 1526 | ||
| XM_005265794.4 | XP_005265851.1 | 12, 13 | 1696 | |||
| XM_005265795.4 | XP_005265852.1 | 12, 13 | 1701 | |||
| GenBank mRNA | AF037195.1 | - | 8, 9 | 958 | ||
| AK098150.1 | - | 5, 6 | 1451 | |||
| AK123382.1 | - | 8, 9 | 870 | |||
| AY987041.1 | - | 8, 9 | 888 | |||
| BC014094.2 | - | 12, 13 | 1496 | 4547415 | ||

RGS14
Genomic Map