siRNA Details
| Gene Symbol: Stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183262.3 | NP_899085.3 | 3 | 1441 | ||
| GenBank mRNA | AK006778.1 | - | 3 | 1206 | ||
| BC047277.1 | - | 2 | 961 | |||
| BC138904.1 | - | 3 | 1348 | 8861926 | ||
| BC138905.1 | - | 3 | 1345 | 8861927 | ||

Stk35
Genomic Map