siRNA Details
| Gene Symbol: Wfdc15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183271.2 | NP_899094.1 | 3 | 352 | ||
| GenBank mRNA | AK006775.1 | - | 3 | 346 | ||
| AK018854.1 | - | 3 | 346 | |||
| AK018888.1 | - | 3 | 353 | |||
| AK018909.1 | - | 3 | 324 | |||
| BC100423.1 | - | 3 | 331 | 30919644 | ||

Wfdc15a
Genomic Map