siRNA Details
| Gene Symbol: Tmem45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183281.2 | NP_899104.2 | 5 | 645 | ||
| GenBank mRNA | AK009173.1 | - | 5 | 653 | ||
| AK036849.1 | - | 4 | 2190 | |||
| BC150914.1 | - | 4 | 798 | |||
| BC150924.1 | - | 4 | 798 | |||
| BC172084.1 | - | 4 | 798 | 9054370 | ||
| CN831346.1 | - | 2 | 58 | |||

Tmem45a2
Genomic Map