siRNA Details
| Gene Symbol: Dhrs13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183286.2 | NP_899109.2 | 4 | 555 | ||
| GenBank mRNA | AK011939.1 | - | 4 | 530 | ||
| AK046465.1 | - | 3 | 602 | |||
| BB637905.1 | - | 4 | 555 | |||
| BC115881.1 | - | 3 | 400 | 40088339 | ||

Dhrs13
Genomic Map