siRNA Details
| Gene Symbol: Tmem140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197986.2 | NP_932103.1 | 2 | 2423 | ||
| GenBank mRNA | AK030532.1 | - | 2 | 2424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem140| Gene Symbol: Tmem140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197986.2 | NP_932103.1 | 2 | 2423 | ||
| GenBank mRNA | AK030532.1 | - | 2 | 2424 | ||