siRNA Details
| Gene Symbol: Emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197991.2 | NP_932108.2 | 2, 3 | 323 | ||
| GenBank mRNA | AK009801.1 | - | 2, 3 | 235 | ||
| AK150499.1 | - | 2, 3 | 228 | |||
| AK160274.1 | - | 2, 3 | 238 | |||
| AK169010.1 | - | 2, 3 | 274 | |||
| AK171657.1 | - | 2, 3 | 237 | |||
| AY761096.1 | - | 2, 3 | 216 | |||
| AY761098.1 | - | 2, 3 | 216 | |||
| BC020179.1 | - | 2, 3 | 218 | 4913500 | ||
| BY271382.1 | - | 2, 3 | 330 | |||

Emc10
Genomic Map