siRNA Details
| Gene Symbol: Smarcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114096.1 | NP_001107568.1 | 2 | 220 | ||
| NM_001114097.1 | NP_001107569.1 | 2 | 220 | |||
| NM_198160.2 | NP_937803.1 | 2 | 220 | |||
| GenBank mRNA | AK089461.1 | - | 2 | 155 | ||
| AK146971.1 | - | 2 | 149 | |||
| AK170530.1 | - | 2 | 155 | |||
| BC040363.1 | - | 2 | 135 | |||
| BC058720.1 | - | 2 | 162 | 5705438 | ||
| CB248220.1 | - | 2 | 220 | |||

Smarcc2
Genomic Map