siRNA Details
| Gene Symbol: Inf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198411.2 | NP_940803.2 | 17 | 2718 | ||
| GenBank mRNA | BC024095.1 | - | 10 | 860 | ||
| BC060610.1 | - | 10 | 969 | 6416724 | ||
| BC099931.1 | - | 10 | 866 | 30063439 | ||
| BC115422.1 | - | 10 | 939 | |||
| BC115423.1 | - | 10 | 939 | |||
| DQ834374.1 | - | 17 | 2726 | |||

Inf2
Genomic Map