siRNA Details
| Gene Symbol: Htra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019752.3 | NP_062726.3 | 3 | 1026 | ||
| GenBank mRNA | AF164513.1 | - | 3 | 905 | ||
| AF175324.1 | - | 3 | 922 | |||
| AK011536.1 | - | 1832 | ||||
| AK146338.1 | - | 3 | 917 | |||
| AK159188.1 | - | 3 | 846 | |||
| BC033276.1 | - | 3 | 886 | |||
| BC049880.1 | - | 3 | 1026 | 5321861 | ||

Htra2
Genomic Map