siRNA Details
| Gene Symbol: Acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019800.4 | NP_062774.2 | 2 | 682 | ||
| GenBank mRNA | AB030038.1 | - | 2 | 675 | ||
| AF216223.1 | - | 2 | 159 | |||
| AK002783.1 | - | 2 | 596 | |||
| AK077729.1 | - | 2 | 670 | |||
| AK160271.1 | - | 2 | 601 | |||
| AK165356.1 | - | 2 | 683 | |||
| AK171757.1 | - | 2 | 597 | |||
| BC047276.1 | - | 2 | 422 | 6487551 | ||

Acp6
Genomic Map