siRNA Details
| Gene Symbol: Slurp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020519.1 | NP_065265.1 | 2 | 181 | ||
| GenBank mRNA | AK003904.1 | - | 2 | 179 | ||
| AK029079.1 | - | 189 | ||||
| BC111105.1 | - | 2 | 180 | 40052992 | ||
| BC125244.1 | - | 2 | 180 | 40053004 | ||

Slurp1
Genomic Map