siRNA Details
| Gene Symbol: Slc25a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020520.4 | NP_065266.1 | 9 | 1555 | ||
| GenBank mRNA | AK075624.1 | - | 9 | 1556 | ||
| AK152436.1 | - | 9 | 1526 | |||
| BC029733.1 | - | 9 | 1552 | 4019406 | ||
| BC052871.1 | - | 9 | 1478 | 30032324 | ||

Slc25a20
Genomic Map