siRNA Details
| Gene Symbol: Smpdl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020561.2 | NP_065586.3 | 3 | 518 | ||
| GenBank mRNA | AK002455.1 | - | 3 | 457 | ||
| AK151683.1 | - | 3 | 404 | |||
| AK152282.1 | - | 3 | 419 | |||
| AK185060.1 | - | 2 | 264 | |||
| AK194580.1 | - | 2 | 264 | |||
| AK205531.1 | - | 2 | 265 | |||
| BC139288.1 | - | 3 | 410 | 8862310 | ||
| BC139289.1 | - | 3 | 410 | 8862311 | ||
| Y08135.1 | - | 3 | 472 | |||

Smpdl3a
Genomic Map