siRNA Details
| Gene Symbol: March7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020575.2 | NP_065600.1 | 6, 7 | 1943 | ||
| GenBank mRNA | AF155739.1 | - | 6, 7 | 1943 | ||
| AK034109.1 | - | 1933 | ||||
| AK045853.1 | - | 1, 2 | 32 | |||
| AK143177.1 | - | 6, 7 | 1965 | |||
| AK166666.1 | - | 6, 7 | 1950 | |||
| AK167246.1 | - | 6, 7 | 1925 | |||
| AK168555.1 | - | 6, 7 | 1946 | |||
| BC025029.1 | - | 6, 7 | 1907 | 4935124 | ||

March7
Genomic Map